4-Bromo-2-methyl-2',4'-bis(trifluoromethyl)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

56865-500MG 56865-1G 56865-5G 56865-25G
77329-774EA 206 USD
77329-774 77353-724 77366-054 77387-872
4-Bromo-2-methyl-2',4'-bis(trifluoromethyl)-1,1'-biphenyl ≥95%
4-Bromo-2-methyl-2',4'-bis(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₉BrF₆
MW: 383.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530552
CAS Number: 2734773-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR