4-Bromo-2-methyl-2'-(trifluoromethyl)-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

56863-500MG 56863-1G 56863-5G 56863-25G
77329-770EA 184.37 USD
77329-770 77347-162 77366-050 77387-868
4-Bromo-2-methyl-2'-(trifluoromethyl)-1,1'-biphenyl ≥97%
4-Bromo-2-methyl-2'-(trifluoromethyl)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrF₃
MW: 315.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530550
CAS Number: 2734773-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR