4'-Bromo-2,2',3-trimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

56861-500MG 56861-1G 56861-5G 56861-25G
77329-768EA 158.62 USD
77329-768 77345-738 77366-048 77387-866
4'-Bromo-2,2',3-trimethyl-1,1'-biphenyl ≥97%
4'-Bromo-2,2',3-trimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₅Br
MW: 275.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530548
CAS Number: 2734773-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR