1,3-Dichloro-4-iodo-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

56839-500MG 56839-1G 56839-5G 56839-25G
77329-764EA 141.11 USD
77329-764 77344-842 77366-046 77387-864
1,3-Dichloro-4-iodo-2-methylbenzene ≥95%
1,3-Dichloro-4-iodo-2-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂I
Storage Temperature: Ambient
MDL Number: MFCD22571085
CAS Number: 190722-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR