3-Ethyl-4-methoxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

56834-500MG 56834-1G 56834-5G 56834-25G
77329-760EA 204.97 USD
77329-760 77353-710 77366-042 77387-860
3-Ethyl-4-methoxy-5-methylbenzaldehyde ≥97%
3-Ethyl-4-methoxy-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD06247465
CAS Number: 883527-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR