3-Ethyl-4-methoxy-5-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

56833-500MG 56833-1G 56833-5G 56833-25G
77329-758EA 184.37 USD
77329-758 77347-232 77366-040 77387-858
3-Ethyl-4-methoxy-5-methylbenzoic acid ≥97%
3-Ethyl-4-methoxy-5-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD34757954
CAS Number: 2709660-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR