(2,4-Dichloro-3-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

56829-500MG 56829-1G 56829-5G 56829-25G
77329-756EA 173.04 USD
77329-756 77346-432 77366-038 77387-856
(2,4-Dichloro-3-methylphenyl)(methyl)sulfane ≥97%
(2,4-Dichloro-3-methylphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₈H₈Cl₂S
MW: 207.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28787038
CAS Number: 1803780-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR