1-Bromo-2,4-dichloro-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

56828-500MG 56828-1G 56828-5G
77340-312EA 900.22 USD
77340-312 77353-708 77377-942
1-Bromo-2,4-dichloro-5-iodobenzene ≥97%
1-Bromo-2,4-dichloro-5-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂I
Storage Temperature: Ambient
MDL Number: MFCD34564407
CAS Number: 2384720-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR