(5-Bromo-2,4-dichlorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

56827-500MG 56827-1G 56827-5G
77339-036EA 545.9 USD
77339-036 77353-706 77377-940
(5-Bromo-2,4-dichlorophenyl)(methyl)sulfane ≥97%
(5-Bromo-2,4-dichlorophenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂S
MW: 271.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564406
CAS Number: 2168080-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR