1-(3-Chloro-2,4-difluorophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

56815-500MG 56815-1G 56815-5G 56815-25G
77329-754EA 173.04 USD
77329-754 77346-450 77366-036 77387-854
1-(3-Chloro-2,4-difluorophenyl)ethanol ≥97%
1-(3-Chloro-2,4-difluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₇ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD34564405
CAS Number: 1894899-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR