2-Bromo-4-ethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

56792-500MG 56792-1G 56792-5G
77329-752EA 347.11 USD
77329-752 77353-704 77377-936
2-Bromo-4-ethoxybenzoic acid ≥97%
2-Bromo-4-ethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23864720
CAS Number: 1427080-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR