2-Bromo-4-ethoxy-1-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

56789-500MG 56789-1G 56789-5G
77329-750EA 342.99 USD
77329-750 77353-700 77377-932
2-Bromo-4-ethoxy-1-iodobenzene ≥97%
2-Bromo-4-ethoxy-1-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrIO
MW: 326.957 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24115729
CAS Number: 1533183-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR