(3-Chloro-2-fluoro-6-iodophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

56787-500MG 56787-1G 56787-5G
77339-370EA 582.98 USD
77339-370 77353-698 77377-930
(3-Chloro-2-fluoro-6-iodophenyl)methanol ≥97%
(3-Chloro-2-fluoro-6-iodophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅ClFIO
MW: 286.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564404
CAS Number: 2386611-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR