(3-Chloro-2-fluoro-6-iodophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

56782-1G 56782-5G 56782-25G
77341-796EA 104.03 USD
77341-796 77366-032 77377-924
(3-Chloro-2-fluoro-6-iodophenyl)(methyl)sulfane ≥97%
(3-Chloro-2-fluoro-6-iodophenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₇H₅ClFIS
MW: 302.535 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564402
CAS Number: 2706922-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR