6-Bromo-3-fluoro-2-iodophenol ≥97%

Supplier: AOBChem USA

Warning

56780-500MG 56780-1G 56780-5G
77337-442EA 414.06 USD
77337-442 77353-692 77377-922
6-Bromo-3-fluoro-2-iodophenol ≥97%
6-Bromo-3-fluoro-2-iodophenol

Precaución: for Research Use Only

Formula: C₆H₃BrFIO
Storage Temperature: Ambient
MDL Number: MFCD28739595
CAS Number: 1805191-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR