1,3-Difluoro-2-iodo-4-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

56766-500MG 56766-1G 56766-5G 56766-25G
77329-748EA 153.47 USD
77329-748 77345-508 77366-030 77387-852
1,3-Difluoro-2-iodo-4-isopropoxybenzene ≥97%
1,3-Difluoro-2-iodo-4-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉F₂IO
MW: 298.068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564401
CAS Number: 2734778-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR