1,3-Difluoro-4-isopropoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

56765-500MG 56765-1G 56765-5G 56765-25G
77329-746EA 229.69 USD
77329-746 77353-690 77366-028 77387-850
1,3-Difluoro-4-isopropoxy-2-methylbenzene ≥97%
1,3-Difluoro-4-isopropoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₂O
MW: 186.198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23114254
CAS Number: 1395037-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR