(3-(Benzyloxy)-2,6-difluorophenyl)(methyl)sulphane ≥97%

Supplier: AOBChem USA

Warning

56764-500MG 56764-1G 56764-5G 56764-25G
77329-744EA 118.45 USD
77329-744 77343-560 77366-026 77377-920
(3-(Benzyloxy)-2,6-difluorophenyl)(methyl)sulphane ≥97%
(3-(Benzyloxy)-2,6-difluorophenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂OS
Storage Temperature: Ambient
MDL Number: MFCD34757932
CAS Number: 2764730-59-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR