(3-Ethoxy-2,6-difluorophenyl)(methyl)sulphane ≥97%

Supplier: AOBChem USA

Warning

56761-500MG 56761-1G 56761-5G 56761-25G
77329-742EA 114.33 USD
77329-742 77343-364 77366-024 77377-918
(3-Ethoxy-2,6-difluorophenyl)(methyl)sulphane ≥97%
(3-Ethoxy-2,6-difluorophenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₉H₁₀F₂OS
Storage Temperature: Ambient
MDL Number: MFCD34564400
CAS Number: 2386274-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR