1-Ethoxy-2,4-difluoro-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

56760-500MG 56760-1G 56760-5G 56760-25G
77329-740EA 120.51 USD
77329-740 77343-752 77366-022 77377-916
1-Ethoxy-2,4-difluoro-3-iodobenzene ≥97%
1-Ethoxy-2,4-difluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₇F₂IO
MW: 284.042 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564399
CAS Number: 2091906-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR