1-Ethoxy-2,4-difluoro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

56759-500MG 56759-1G 56759-5G
77338-188EA 468.65 USD
77338-188 77353-688 77377-914
1-Ethoxy-2,4-difluoro-3-methylbenzene ≥97%
1-Ethoxy-2,4-difluoro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O
Storage Temperature: Ambient
MDL Number: MFCD28788832
CAS Number: 1806313-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR