(4-Ethoxy-3-methoxyphenyl)(methyl)sulphane ≥97%

Supplier: AOBChem USA

Warning

56741-500MG 56741-1G 56741-5G 56741-25G
77329-736EA 193.64 USD
77329-736 77353-684 77366-018 77387-846
(4-Ethoxy-3-methoxyphenyl)(methyl)sulphane ≥97%
(4-Ethoxy-3-methoxyphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂S
Storage Temperature: Ambient
MDL Number: MFCD34757919
CAS Number: 2901086-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR