1-(4-Ethoxy-3-methoxyphenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

56740-500MG 56740-1G 56740-5G 56740-25G
77329-734EA 243.08 USD
77329-734 77353-682 77366-016 77387-844
1-(4-Ethoxy-3-methoxyphenyl)ethanol ≥97%
1-(4-Ethoxy-3-methoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD11053276
CAS Number: 1048917-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR