1-(4-Ethoxy-3-methoxyphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

56739-500MG 56739-1G 56739-5G 56739-25G
77329-732EA 160.68 USD
77329-732 77345-814 77366-014 77387-842
1-(4-Ethoxy-3-methoxyphenyl)ethanone ≥97%
1-(4-Ethoxy-3-methoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02934033
CAS Number: 75665-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR