2-(3,4-Dimethoxyphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

56736-500MG 56736-1G 56736-5G 56736-25G
77329-730EA 261.62 USD
77329-730 77353-680 77366-012 77387-840
2-(3,4-Dimethoxyphenyl)propan-2-ol ≥95%
2-(3,4-Dimethoxyphenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₃
MW: 196.243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222266
CAS Number: 74385-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR