4-Fluoro-3-(methylsulfinyl)phenol ≥95%

Supplier: AOBChem USA

Warning

56728-500MG 56728-1G 56728-5G 56728-25G
77329-728EA 141.11 USD
77329-728 77344-840 77366-008 77387-838
4-Fluoro-3-(methylsulfinyl)phenol ≥95%
4-Fluoro-3-(methylsulphinyl)phenol

Precaución: for Research Use Only

Formula: C₇H₇FO₂S
MW: 174.193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530542
CAS Number: 2734777-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR