1-Ethoxy-2-fluoro-5-iodo-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

56709-500MG 56709-1G 56709-25G 56709-5G
77329-726EA 294.58 USD
77329-726 77353-676 77387-836 77377-908
1-Ethoxy-2-fluoro-5-iodo-3-methylbenzene ≥97%
1-Ethoxy-2-fluoro-5-iodo-3-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀FIO
MW: 280.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757906
CAS Number: 2090745-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR