(3-Ethoxy-4-fluoro-5-methylphenyl)(methyl)sulphane ≥97%

Supplier: AOBChem USA

Warning

56708-500MG 56708-1G 56708-5G 56708-25G
77329-724EA 278.1 USD
77329-724 77353-674 77366-006 77387-834
(3-Ethoxy-4-fluoro-5-methylphenyl)(methyl)sulphane ≥97%
(3-Ethoxy-4-fluoro-5-methylphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₁₀H₁₃FOS
Storage Temperature: Ambient
MDL Number: MFCD34757905
CAS Number: 2711575-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR