2-(3-Ethoxy-4-fluoro-5-methylphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

56699-500MG 56699-1G 56699-25G 56699-5G
77329-720EA 297.67 USD
77329-720 77353-670 77387-832 77377-904
2-(3-Ethoxy-4-fluoro-5-methylphenyl)propan-2-ol ≥97%
2-(3-Ethoxy-4-fluoro-5-methylphenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₂H₁₇FO₂
Storage Temperature: Ambient
MDL Number: MFCD34757900
CAS Number: 2755719-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR