2-Fluoro-5-iodo-3-methylphenol ≥97%

Supplier: AOBChem USA

Warning

56686-500MG 56686-1G 56686-5G 56686-25G
77329-718EA 150.38 USD
77329-718 77345-358 77366-004 77387-830
2-Fluoro-5-iodo-3-methylphenol ≥97%
2-Fluoro-5-iodo-3-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆FIO
Storage Temperature: Ambient
MDL Number: MFCD34757887
CAS Number: 2384189-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR