2-(2-Fluoro-5-iodo-3-methylphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

56684-500MG 56684-1G 56684-5G 56684-25G
77329-716EA 256.47 USD
77329-716 77353-664 77366-002 77387-828
2-(2-Fluoro-5-iodo-3-methylphenyl)propan-2-ol ≥97%
2-(2-Fluoro-5-iodo-3-methylphenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₂FIO
MW: 294.105 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757885
CAS Number: 2757731-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR