1-(2-Fluoro-5-iodo-3-methylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

56683-500MG 56683-1G 56683-5G 56683-25G
77329-714EA 163.77 USD
77329-714 77346-116 77366-000 77387-826
1-(2-Fluoro-5-iodo-3-methylphenyl)ethanone ≥97%
1-(2-Fluoro-5-iodo-3-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈FIO
Storage Temperature: Ambient
MDL Number: MFCD34757884
CAS Number: 2384686-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR