1-(2-Fluoro-3-(methylthio)-5-(trifluoromethyl)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

56677-500MG 56677-1G 56677-5G 56677-25G
77329-712EA 233.81 USD
77329-712 77353-662 77365-998 77387-824
1-(2-Fluoro-3-(methylthio)-5-(trifluoromethyl)phenyl)ethanone ≥97%
1-(2-Fluoro-3-(methylthio)-5-(trifluoromethyl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₈F₄OS
MW: 252.229 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530541
CAS Number: 2734779-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR