(2-Fluoro-5-(trifluoromethyl)-1,3-phenylene)bis(methylsulfane) ≥97%
Supplier: AOBChem USA
|
Warning
|
56676-500MG
56676-1G
56676-5G
56676-25G
77329-710EA
82.4
USD
77329-710
77342-154
77365-996
77377-898
(2-Fluoro-5-(trifluoromethyl)-1,3-phenylene)bis(methylsulfane) ≥97%
(2-Fluoro-5-(trifluoromethyl)-1,3-phenylene)bis(methylsulphane)
Precaución: for Research Use Only
Formula:
C₉H₈F₄S₂ MW: 256.283 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34530540 CAS Number: 2734779-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...