Methyl 2-fluoro-3-(methylthio)-5-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

56673-500MG 56673-1G 56673-5G 56673-25G
77329-708EA 167.89 USD
77329-708 77346-280 77365-994 77387-822
Methyl 2-fluoro-3-(methylthio)-5-(trifluoromethyl)benzoate ≥97%
Methyl 2-fluoro-3-(methylthio)-5-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₈F₄O₂S
Storage Temperature: Ambient
MDL Number: MFCD34757880
CAS Number: 2901098-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR