2-Fluoro-3-(methylthio)-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

56672-500MG 56672-1G 56672-5G 56672-25G
77329-706EA 142.14 USD
77329-706 77344-972 77365-992 77387-820
2-Fluoro-3-(methylthio)-5-(trifluoromethyl)benzoic acid ≥97%
2-Fluoro-3-(methylthio)-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₆F₄O₂S
MW: 254.201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530538
CAS Number: 2734778-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR