2,4-Difluoro-5-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

56653-500MG 56653-1G 56653-5G 56653-25G
77329-704EA 126.69 USD
77329-704 77344-212 77365-990 77377-894
2,4-Difluoro-5-methoxyphenol ≥97%
2,4-Difluoro-5-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆F₂O₂
MW: 160.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28789016
CAS Number: 1803822-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR