(2,4-Difluoro-5-methoxyphenyl)(methyl)sulphane ≥97%

Supplier: AOBChem USA

Warning

56649-500MG 56649-1G 56649-5G
77336-424EA 387.28 USD
77336-424 77353-656 77377-890
(2,4-Difluoro-5-methoxyphenyl)(methyl)sulphane ≥97%
(2,4-Difluoro-5-methoxyphenyl)(methyl)sulphane

Precaución: for Research Use Only

Formula: C₈H₈F₂OS
Storage Temperature: Ambient
MDL Number: MFCD34564398
CAS Number: 2383386-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR