(4-Iodo-3-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

56640-500MG 56640-1G 56640-5G 56640-25G
77329-702EA 235.87 USD
77329-702 77353-654 77365-988 77387-818
(4-Iodo-3-methylphenyl)methanol ≥97%
(4-Iodo-3-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₉IO
Storage Temperature: Ambient
MDL Number: MFCD11036281
CAS Number: 959632-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR