(2-Iodo-5-methoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

56630-500MG 56630-1G 56630-5G 56630-25G
77329-700EA 99.91 USD
77329-700 77342-740 77365-986 77377-888
(2-Iodo-5-methoxyphenyl)methanol ≥95%
(2-Iodo-5-methoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₉IO₂
Storage Temperature: Ambient
MDL Number: MFCD09909620
CAS Number: 139557-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR