(2-Iodo-5-methylphenyl)methanol ≥97%
Supplier: AOBChem USA
|
Warning
|
56606-1G
56606-5G
56606-25G
77342-094EA
113.3
USD
77342-094
77365-984
77377-884
(2-Iodo-5-methylphenyl)methanol ≥97%
(2-Iodo-5-methylphenyl)methanol
Precaución: for Research Use Only
Formula:
C₈H₉IO MW: 248.06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18397811 CAS Number: 220991-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...