Ethyl 3-(3,5-bis(trifluoromethyl)phenyl)-3-hydroxypropanoate ≥97%

Supplier: AOBChem USA

Warning

56579-500MG 56579-1G 56579-5G 56579-25G
77329-698EA 95.79 USD
77329-698 77342-562 77365-980 77377-880
Ethyl 3-(3,5-bis(trifluoromethyl)phenyl)-3-hydroxypropanoate ≥97%
Ethyl 3-(3,5-bis(trifluoromethyl)phenyl)-3-hydroxypropanoate

Precaución: for Research Use Only

Formula: C₁₃H₁₂F₆O₃
MW: 330.223 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564389
CAS Number: 2734776-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR