Methyl 3-(2-bromo-5-methoxyphenyl)-3-hydroxypropanoate ≥95%

Supplier: AOBChem USA

Warning

56575-1G 56575-5G 56575-25G
77341-164EA 72.1 USD
77341-164 77365-978 77377-878
Methyl 3-(2-bromo-5-methoxyphenyl)-3-hydroxypropanoate ≥95%
Methyl 3-(2-bromo-5-methoxyphenyl)-3-hydroxypropanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₄
MW: 289.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530535
CAS Number: 1621506-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR