5-Bromo-4-fluoro-2-methoxyphenyl acetate ≥97%

Supplier: AOBChem USA

Warning

56546-1G 56546-5G 56546-25G
77340-744EA 60.77 USD
77340-744 77365-972 77377-872
5-Bromo-4-fluoro-2-methoxyphenyl acetate ≥97%
5-Bromo-4-fluoro-2-methoxyphenyl acetate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD34530530
CAS Number: 2734773-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR