Methyl 3-(2,6-difluorophenyl)-3-hydroxypropanoate ≥97%

Supplier: AOBChem USA

Warning

56519-500MG 56519-1G 56519-5G
77337-624EA 431.57 USD
77337-624 77353-650 77377-870
Methyl 3-(2,6-difluorophenyl)-3-hydroxypropanoate ≥97%
Methyl 3-(2,6-difluorophenyl)-3-hydroxypropanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₃
MW: 216.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564368
CAS Number: 2229575-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR