1-Bromo-3-fluoro-2-(methoxymethoxy)-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

56435-500MG 56435-1G 56435-5G 56435-25G
77329-690EA 126.69 USD
77329-690 77344-142 77365-966 77377-858
1-Bromo-3-fluoro-2-(methoxymethoxy)-4-methylbenzene ≥97%
1-Bromo-3-fluoro-2-(methoxymethoxy)-4-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34530524
CAS Number: 1934816-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR