Methyl 3-bromo-5-fluoro-2-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

56427-500MG 56427-1G 56427-5G
77329-686EA 319.3 USD
77329-686 77353-636 77377-854
Methyl 3-bromo-5-fluoro-2-methoxybenzoate ≥95%
Methyl 3-bromo-5-fluoro-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₃
Storage Temperature: Ambient
MDL Number: MFCD29922181
CAS Number: 1956380-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR