3-Bromo-5-fluoro-2-methoxy-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

56417-500MG 56417-1G 56417-5G 56417-25G
77329-684EA 204.97 USD
77329-684 77353-632 77365-964 77387-814
3-Bromo-5-fluoro-2-methoxy-N-methylbenzamide ≥97%
3-Bromo-5-fluoro-2-methoxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD34757804
CAS Number: 2755720-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR