3-Bromo-5-fluoro-2-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

56416-500MG 56416-1G 56416-5G 56416-25G
77329-682EA 258.53 USD
77329-682 77353-630 77365-962 77387-812
3-Bromo-5-fluoro-2-methoxybenzamide ≥97%
3-Bromo-5-fluoro-2-methoxybenzamide

Precaución: for Research Use Only

Formula: C₈H₇BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD29920625
CAS Number: 1956327-35-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR