(5-Chloro-2-fluoro-3-iodophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

56409-500MG 56409-1G 56409-5G 56409-25G
77329-680EA 212.18 USD
77329-680 77353-628 77365-960 77387-810
(5-Chloro-2-fluoro-3-iodophenyl)methanol ≥97%
(5-Chloro-2-fluoro-3-iodophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅ClFIO
MW: 286.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757803
CAS Number: 2383985-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR